Learn More
Medchemexpress LLC Lidocaine impurity 1 | 39084-88-3 | 99.9% | 232.11 | 50 MG

Supplier: Medchemexpress LLC HY4400950MG
Lidocaine impurity 1 is an impurity of Lidocaine. It is for research use only and not sold to patients. The appearance is solid, and the formula is C10H11Cl2NO.
- Appearance: Solid
- Formula: C10H11Cl2NO
- Shipping: Room temperature in continental US; may vary elsewhere
- Storage (powder): -20°C for 3 years, 4°C for 2 years
- Storage (in solvent): -80°C for 6 months, -20°C for 1 month
- Usage: For research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.