Learn More
Medchemexpress LLC Lidocaine N-oxide | 2903-45-9 | MFCD22125322 | 99.9% | 250.34 | C14H22N2O2 | 10MG

Supplier: Medchemexpress LLC HY12409810MG
Lignocaine N-oxide (lidocaine N-oxide) is a drug metabolite used as a reference standard in biochemical and pharmacological research. It is supplied as a powder with characterized solubility and storage recommendations for laboratory use.
- CAS number 2903-45-9 for unambiguous identification.
- Chemical formula C14H22N2O2 and molecular weight 250.34 g/mol.
- Purity 99.9% (reported by supplier) suitable for analytical applications.
- Appearance: powder; solubility: DMSO 100 mg/mL (requires ultrasonic), aqueous formulations achieving ≥5 mg/mL.
- Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
- Available in mg-scale solid packs and 10 mM solution formats for research use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.