Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Lidocaine N-oxide | 2903-45-9 | MFCD22125322 | 99.9% | 250.34 g/mol | C14H22N2O2 | 5MG
SDP

Supplier:  Medchemexpress LLC HY1240985MG

Encompass_Preferred

Lignocaine N-oxide (lidocaine N-oxide) is the N-oxide metabolite of lidocaine provided as a research-grade chemical standard for metabolic and biochemical studies. It is used as a reference material to investigate lidocaine metabolism and related pathways in vitro and in microsomal systems.

  • N-oxide metabolite of lidocaine used to study lidocaine metabolism.
  • Molecular formula C14H22N2O2; molecular weight 250.34 g/mol.
  • Purity 99.92% (manufacturer reported).
  • Solid appearance for standard laboratory handling.
  • Supplied in small research quantities (for example, 5 mg).

Catalog No. 50-002-37967


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.