Learn More
Medchemexpress LLC Lidocaine (Standard) | 137-58-6 | 100.0% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB0185R50MG
Encompass_Preferred
Lidocaine (Standard) is the analytical standard of Lidocaine, intended for research and analytical applications. It inhibits sodium channels involving complex voltage and using dependence, and decreases growth, migration and invasion of gastric carcinoma cells via up-regulating miR-145 expression and further inactivation of MEK/ERK and NF-κB signaling pathways. It is an amide derivative and has potential for the research of ventricular arrhythmia.
  • Analytical standard for research and analytical applications
  • Inhibits sodium channels
  • Decreases growth, migration, and invasion of gastric carcinoma cells
  • Potential for research of ventricular arrhythmia
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications
Catalog No. 50-004-07343