Learn More
Medchemexpress LLC Lidocaine (Standard) | 137-58-6 | 99.9% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB0185R
Encompass_Preferred
Lidocaine (Standard) is the analytical standard of Lidocaine, intended for research and analytical applications. Lidocaine (Lignocaine) inhibits sodium channels involving complex voltage and using dependence. It decreases growth, migration, and invasion of gastric carcinoma cells by up-regulating miR-145 expression and further inactivating MEK/ERK and NF-κB signaling pathways. Lidocaine is an amide derivative and has potential for the research of ventricular arrhythmia.
  • The compound is the grade of analytical standard, which is the reference standard supplied assay.
  • It is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
Catalog No. 50-004-50609