Learn More
Research Products International Corp Silicone Lids, Mixed Size Set, PNK, 3/pk
SDP
Supplier:  Research Products International Corp 212303P
Encompass
Kimble Silicone Lids are made of stretchable, chemically and heat resistant silicone. Reusable, versatile lids fit on beakers, flasks, bottles, cylinders, plastic and glass. Adaptable lids conform to various container spouts and thread finishes. Colored lids stay secure and defend against spills, leakage and reduce environmental impact over foil or film. Ideal for color coding projects, samples and compatible with markers to enhance sample identification. Silicone lids are dishwasher safe and remain pliable over a wide temperature range. Recommended usable temperature range is -40° to +180°C.
Versatile safety lids
Reusable and autoclavable
Bright Colors
Catalog No. 50-222-9656