Learn More
Medchemexpress LLC HY-N0484 5mg Medchemexpress, Liensinine CAS:2586-96-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN04845MG
Encompass_Preferred
Medchemexpress, HY-N0484 5mg Liensinine CAS:2586-96-1 Liensinine is an autophagy/mitophagy inhibitor. Liensinine, a major isoquinoline alkaloid, extracted from the seed embryo of Nelumbo nucifera Gaertn, has a wide range of biological activities, including anti-arrhythmias, anti-hypertension, anti-pulmonary fibrosis, relaxation on vascular smooth muscle, etc. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0277