Learn More
Medchemexpress LLC LIF Human HEK293 100ug
SDP
Supplier:  Medchemexpress LLC HYP73276100UG
Encompass_Preferred
LIF protein induces terminal differentiation in leukemic cells promoting hematopoietic and neuronal cell differentiation and stimulating acute-phase protein synthesis in hepatocytes Its diverse activities highlight its role in influencing cellular differentiation across various cell types and physiological contexts LIF Protein Human (HEK293) is the recombinant human-derived LIF protein expressed by HEK293 with tag free
Catalog No. 50-001-80275