Learn More
TARGETMOL CHEMICALS INC LIFARIZINE FA 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T27830L5MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Lifarizine FA is a sodium channel blocker used in the treatment of neurological disorders and cardiovascular diseases study of stroke.
Catalog No. 50-258-0567