Learn More
Cayman Chemical LIfItegrast 10mg
SDP
Supplier:  Cayman Chemical 2258810 MG
Encompass_Preferred
An inhibitor of the LFA-1/ICAM-1 interaction; binds to LFA-1 and blocks the adhesion of Jurkat T cells or HuT 78 cells to ICAM-1 (IC50s = 2.98 and 9 nM, respectively) and inhibits secretion of various cytokines from these cells; increases tear production in dogs with idiopathic keratoconjunctivitis sicca; topical application inhibits LFA-1-dependent neutrophil recruitment to the eye in mouse models of contact lens-associated corneal inflammation induced by corneal abrasion and tobramycin-killed bacteria
Catalog No. 50-298-6723