Learn More
MSC Liftex Endless Round Sling, 5300 lb Vertical Capacity, 10 ft Length, Polyester
Supplier: MSC 76036110
This endless round sling is designed for high-capacity lifting applications. Made from 100% polyester fiber with a double polyester woven jacket, it offers a 5300 lb vertical capacity and measures 10 ft in length. Its green color signifies its capacity rating.
- Strong grip on loads
- Easy handling and storage
- Tighter choke with an easier release
- Risk reduction of back and hand injuries
- Reduced cost with less total rigging weight
- Tough, resilient tag for quick and easy identification