Learn More
Electron Microscopy Sciences Lifterslip™ Hybridization mSeries Slide, 25 x 25 mm, 23 mcL
SDP
Supplier:  Electron Microscopy Sciences 7218325
Encompass_Preferred
The mSeries LifterSlip Solves the problem of hybridization non-uniformity. mSeries LifterSlip are manufactured from 1 mm thick glass specially drawn for flatness and superior surface quality. The 1 mm thickness has increased stiffness that reduces variation in hybridization fluid volume. Precise ink printing provides an innovative raised edge design for consistent dispersal and easy injection of hybridization solution under the prepositioned lifterslip.
Size: 25 x 25 mm
Volume: 23 µl
Catalog No. 50-189-9757