Learn More
Cayman Chemical LIG4 BRCT domaInshumnrec 100ug
SDP
Supplier:  Cayman Chemical 14169100 UG
Encompass_Preferred
Source: Recombinant N-terminal GST-tagged protein expressed in E. coli • AA: 1634-1863 • MW: 57.6 kDa
Catalog No. 50-295-5475