Learn More
Electron Microscopy Sciences Light Green SF Yellowish, Certified, C.I. DcL-30 10 GR

Supplier: Electron Microscopy Sciences 17920
(Acid Green 5; Light Green 2G, S or 2GN; Acid Green F; Fast Acid Green N)
CAS #5141-20-8 C₃₇H₃₄N₂Na₂O₉S₃ F.W. 792.86
Solubility: 20.35% Water; 4.0% Alcohol; 5.25% Cellosolve; 12.0% Glycol; 0.0% Xylene.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.