Learn More
Perkin Elmer US LLC Light Shield for the Fiber Optic Dip Probe
Supplier:  Perkin Elmer US LLC N1016074
Encompass_Preferred
The Fiber Optic Dip Probe is compatible with the LAMBDA 20/20Bio/40/40Bio/25/35/45 650/750/850/850+/950/1050/1050+ spectrometers. It requires, but does not include: Transfer-Optic Mono-Fibers (B0509546 or L6020290) a Light Shield (N1016074) a Probe Tip (N1016073, N1016072, or N1016071)
Catalog No. 50-210-4148