Learn More
Darwin Chambers Company LITHIUM-IRON BATTERY PACK
SDP
Supplier:  Darwin Chambers Company RBP
Encompass_Preferred
Rechargeable Lithium-Iron Battery Pack for Portable Incubator. Ultra light. One fifth the weight of lead-acid batteries on average. Twice to four times the service life of lead-acid batteries. Super-fast recharge rate. No explosive gasses during charge, no lead, no acid. Environmentally friendly. Short circuit protection. No presence of toxic substances. Includes charging adapter and all necessary cables.
Catalog No. NC0661041