Learn More
ATCC Ligilactobacillus salivarius (Rogosa et al.) Zheng et al. strain NCIB 8816, freeze-dried culture (ATCC 29602)
Supplier:  ATCC 29602
Encompass
Ligilactobacillus salivarius strain NCIB 8816 (ATCC 29602) is a freeze-dried bacterial culture isolated from human saliva and provided for laboratory research. The strain is supplied lyophilized for secure transport and long-term storage and can be revived and cultivated under standard lactobacilli conditions.
  • Freeze-dried preparation for long-term storage.
  • Store refrigerated at 2 °C to 8 °C.
  • Cultivate on MRS agar or broth (lactobacilli medium).
  • Incubate at 37 °C under aerobic conditions.
  • Biosafety level 1 organism suitable for basic laboratory research.
  • Isolated from human saliva; originally obtained in Italy.
Catalog No. NC3803615