Learn More
Medchemexpress LLC Lignan P | 23363-35-1 | C27H30O13 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13666910MG
Encompass_Preferred
Etoposide impurity 1 (Lignan P, CAS 23363-35-1) is an analytical impurity standard used for identification and quantification of etoposide-related impurities. Supplied as a solid reference material, it supports analytical testing, method development, impurity profiling, and quality control. Typical specifications include molecular formula C27H30O13, molecular weight 562.52, and purity >95% (HPLC).
  • Analytical reference for impurity identification and quantitation.
  • Suitable for HPLC, LC-MS, and NMR method development and validation.
  • Supplied as a solid with chemical formula C27H30O13 and MW 562.52.
  • High purity (>95.0% by HPLC) for reliable analytical results.
  • Intended for pharmaceutical impurity profiling and quality control.
Catalog No. 50-002-59956