Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Lignocaine n-oxide (lidocaine n-oxide) | 2903-45-9 | MFCD22125322 | 99.9% | 250.34 g/mol | C14H22N2O2 | 50MG
SDP

Supplier:  Medchemexpress LLC HY12409850MG

Encompass_Preferred

Lignocaine N-oxide (Lidocaine N-oxide) is the N-oxide metabolite of the local anesthetic lidocaine, supplied as a solid research chemical for biochemical and drug metabolism studies. It is characterized by CAS 2903-45-9, formula C14H22N2O2, and a molecular weight of 250.34 g/mol.

  • High purity suitable for analytical research: 99.9%.
  • Empirical data includes molecular weight 250.34 g/mol and formula C14H22N2O2.
  • Physical form is a solid powder, typically supplied in a 50 mg vial.
  • Recommended storage: powder at -20°C for long-term storage; in solvent at -80°C for up to six months.
  • Common applications include drug metabolism studies, analytical standards, and pharmacokinetic research.

Catalog No. 50-002-38426


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.