Learn More
Cayman Chemical LIgnocerIc AcId-d47 5mg
SDP
Supplier:  Cayman Chemical 90033205 MG
Encompass_Preferred
An internal standard for the quantification of lignoceric acid by GC- or LC-MS
Catalog No. 50-289-4307