Learn More
Medchemexpress LLC Lignoceric acid-d9 | 2415226-90-1 | 98.0% | 5 MG

Supplier: Medchemexpress LLC HY121883SS55MG
Lignoceric acid-d9 is the deuterium-labeled form of lignoceric acid (tetracosanoic acid). Supplied as a solid isotope-labeled reference standard, it is intended for use as an internal standard in mass spectrometry, metabolic tracing, and quantitative lipid analysis in research and analytical workflows.
- Deuterium-labeled internal standard for mass spectrometry.
- Molecular weight 377.69 g/mol and formula C24H39D9O2.
- Available in small pack sizes suitable for analytical use.
- Appearance: solid, white to off-white; powder storage recommended at -20°C for long term.
- Pack sizes include 1 mg, 5 mg, and 10 mg.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.