Learn More
Medchemexpress LLC Ligustrazine hydrochloride | 76494-51-4 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN093550MG
Encompass_Preferred
Ligustrazine hydrochloride is a natural product that has shown a protective effect on injured ECV304 cells, with significant increases in NOS and NO formation compared to the model group.
  • Natural product
  • Protective effect on injured ECV304 cells
  • Increases NOS and NO formation
  • White to off-white solid appearance
  • For research use only
  • Optimal storage at 4°C, sealed, away from moisture
  • Soluble in DMSO and H2O (100 mg/mL)
Catalog No. 50-004-16491