Learn More
Cayman Chemical LIgustroflavon 10mg
SDP
Supplier:  Cayman Chemical 2743110 MG
Encompass_Preferred
An apigenin triglycoside with diverse biological activities; increases PTH release from rat primary parathyroid gland cells at 1 µM and inhibits calcium influx in HEK293 cells; increases serum PTH, as well as serum and bone calcium levels, in a streptozotocin-induced mouse model of diabetes with osteoporosis at 5 and 20 mg/kg three times per week; reduces increases in the expression and levels of CaSR in the kidney of diabetic osteoporotic mice; decreases infarct volume in a rat model of ischemic stroke at 10, 30, and 60 mg/kg and decreases the levels of the necroptosis-associated proteins RIPK3 and MLKL in the brain of ischemic rats at 30 mg/kg
Catalog No. 50-293-9179