Learn More
Medchemexpress LLC Ligustroflavone | 260413-62-5 | 724.66 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN05461MG
Encompass_Preferred
Ligustroflavone is an orally active flavonoid compound extracted from Ligustrum lucidum. It acts as a calcium-sensing receptor (CaSR) antagonist, inhibits the RIPK1/RIPK3/MLKL pathway, and downregulates TGF-β/Smad signaling. It regulates calcium metabolism, protects bone tissue, reduces cerebral ischemic injury, and inhibits liver fibrosis.
  • Regulates calcium metabolism
  • Protects bone tissue
  • Reduces cerebral ischemic injury
  • Inhibits liver fibrosis
  • Antagonizes the calcium-sensing receptor (CaSR)
  • Inhibits the RIPK1/RIPK3/MLKL pathway
  • Downregulates TGF-β/Smad signaling
Catalog No. 50-003-88795