Learn More
Medchemexpress LLC Ligustroflavone | 260413-62-5 | 724.66 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN05465MG
Encompass_Preferred
Ligustroflavone is an orally active flavonoid compound extracted from *Ligustrum lucidum*. It acts as a calcium-sensing receptor (CaSR) antagonist, inhibits the RIPK1/RIPK3/MLKL pathway, and downregulates TGF-β/Smad signaling. It plays a role in regulating calcium metabolism, protecting bone tissue, reducing cerebral ischemic injury, and inhibiting liver fibrosis. Ligustroflavone is used in the study of diabetic osteoporosis, ischemic stroke, and liver fibrosis.
  • Orally active flavonoid compound
  • Acts as a calcium-sensing receptor (CaSR) antagonist
  • Inhibits RIPK1/RIPK3/MLKL pathway
  • Downregulates TGF-β/Smad signaling
  • Regulates calcium metabolism and protects bone tissue
  • Reduces cerebral ischemic injury
  • Inhibits liver fibrosis
  • Used in the study of diabetic osteoporosis, ischemic stroke, and liver fibrosis
Catalog No. 50-003-88862