Learn More
Bachem L-Anserine · nitrate
SDP
Supplier:  Bachem 4027810.0100
Encompass_Preferred
L-Anserine · nitrate, H-b-Ala-His(3-Me)-OH · nitrate, 10030-52-1, C10H16N4O3 · HNO3, Mr 303.28, 100mg. Like L-carnosine, L-anserine is a histidine-containing dipeptide found in vertebrate skeletal muscles. Both are potent antioxidants with membrane-stabilizing function.
Catalog No. 50-194-6817