Learn More
Genscript Corporation LILRA1/CD85i/LIR-6Biotin Hi
Supplier:  Genscript Corporation Z04536500
Encompass
LILRA1 also known as CD85i and LIR-6 is an approximately 70 kDa variably glycosylated transmembrane protein that regulates immune cell activation Mature human LILRA1 consists of a 445 amino acid (aa) extracellular domain (ECD) with 4 Ig-like domains a 21 aa transmembrane segment and a 7 aa cytoplasmic tail LILRA1 may act as receptor for class I MHC antigens
Catalog No. 50-000-57993