Learn More
Medchemexpress LLC LILRB2/CD85d/ILT-4 100ug | 100ug
SDP
Supplier:  Medchemexpress LLC HYP77739100UG
Encompass_Preferred
The LILRB2/CD85d/ILT-4 protein is a receptor for class I MHC antigens and can recognize multiple HLA alleles It crucially downregulates the immune response and builds tolerance LILRB2/CD85d/ILT-4 Protein Cynomolgus (HEK293 Fc) is the recombinant cynomolgus-derived LILRB2/CD85d/ILT-4 protein expressed by HEK293 with C-hFc labeled tag
Catalog No. 50-001-87934