Learn More
Medchemexpress LLC Limaprost | 74397-12-9 | 99.9% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB068350MG
Encompass_Preferred
Limaprost (OP1206) is a potent, orally active prostaglandin E1 (PGE1) analogue and vasodilator. It functions by increasing blood flow and inhibiting platelet aggregation, offering potential benefits such as pain relief and antianginal effects. This compound is suitable for research related to ischaemic symptoms.
  • Potent and orally active PGE1 analogue vasodilator.
  • Increases blood flow and inhibits platelet aggregation.
  • Provides pain relief and antianginal effects.
  • Suitable for research on ischaemic symptoms.
  • Inhibits IL-1-mediated induction of nerve growth factor (NGF) in human IVD cells.
  • Demonstrates in vivo efficacy in relieving Argipressin-induced ST depression.
  • Increases coronary blood flow and reduces vascular resistance in animal models.
Catalog No. 50-004-08562