Learn More
Cayman Chemical LImaprost-d3 500ug
SDP
Supplier:  Cayman Chemical 25416500 UG
Encompass_Preferred
An internal standard for the quantification of limaprost by GC- or LC-MS
Catalog No. 50-293-5019