Learn More
Grainger LimitSwitch RollerLeverPlunger 1NO/1NC
Supplier:  Grainger 14H181
Encompass_Preferred
The Dayton Limit Switch Roller Arm Plunger is a limit switch with a roller lever actuator. It has a compact design with a 1/2" roller. The housing is made of zinc alloy with a NEMA 1, 3, 4, 12 and 13 rating for use in various environments. It has SPDT snap action contacts rated for 10A at 125/250VAC. The operating force range is 12-80 oz. The operating temperature range is -13°F to 185°F. The electrical life is 50,000 cycles minimum. This limit switch can be used for conveyors, packaging equipment, machine tools, material handling equipment, and more. It provides position sensing and control.
Catalog No. 50-275-0674