Learn More
Medchemexpress LLC Lin28 inhibitor LI71 enantiomer | 956189-58-5 | 99.7% | 335.40 g/mol | C21H21NO3 | 10MG
SDP
Supplier:  Medchemexpress LLC HY123905A10MG
Encompass_Preferred
LIN28 inhibitor LI71 enantiomer is the less-active stereoisomer of LI71 provided as a high-purity research reagent. It is used to investigate LIN28-mediated regulation of let-7 and related biological interactions, and has reported activity against Plasmodium cold shock protein and tubulin. Supplier stability and storage guidance is available for both powder and solution forms.
Catalog No. 50-002-35710