Learn More
Medchemexpress LLC Lin28 inhibitor LI71 enantiomer | 956189-58-5 | MFCD07403664 | 99.7% | 335.40 | C21H21NO3 | 50MG
SDP
Supplier:  Medchemexpress LLC HY123905A50MG
Encompass_Preferred
LIN28 inhibitor LI71 enantiomer is the less active stereoisomer of LI71 that disrupts LIN28:let-7 binding. Supplied for research use, it is characterized by reported purity, molecular formula, and molecular weight, and is used for biochemical and cellular studies comparing stereoisomer activity.
  • Less active enantiomer useful for stereoisomer comparison.
  • Disrupts LIN28:let-7 interaction in biochemical assays.
  • High reported purity suitable for research applications.
  • Available in multiple pack sizes, including 50 mg.
  • Characterized by CAS number 956189-58-5 and molecular formula C21H21NO3.
Catalog No. 50-002-35939