Learn More
Cayman Chemical LInalol-d3 10mg
SDP
Supplier:  Cayman Chemical 900427310 MG
Encompass_Preferred
An internal standard for the quantification of (±)-linalool by GC- or LC-MS
Catalog No. 50-289-5938