Learn More
Medchemexpress LLC Linalool-d5 | 159592-39-9 | C10H13D5O | 5 MG
Supplier: Medchemexpress LLC HY152052S5MG
Linalool-d5 is a deuterium labeled monoterpene and a competitive NMDA receptor antagonist. It is orally active and can cross the blood-brain barrier, offering a range of protective activities. This compound is intended for research use only, not for patient use.
- Acts as a competitive NMDA receptor antagonist
- Offers a broad spectrum of protective activities including anticancer, antibacterial, and anti-inflammatory effects
- Functions as a tracer for research applications
- Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS