Learn More
Medchemexpress LLC Linalool (Standard) | 78-70-6 | 154.25 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN0368R50MG
Encompass_Preferred
Linalool (Standard) is the analytical standard of Linalool, a natural monoterpene found in essential oils of coriander. This product is intended for research and analytical applications, serving as a reference standard for various assays. Linalool acts as a competitive antagonist of the N-methyl d-aspartate (NMDA) receptor, and also functions as a PPARα ligand.
  • Serves as a reference standard for assays.
  • Used in qualitative research experiments.
  • Used in quantitative research experiments.
  • Used in methodological research experiments.
  • Commonly used with HPLC, GC, and MS.
Catalog No. 50-003-89709