Learn More
Cayman Chemical LInarmycIn B 2.5mg
SDP
Supplier:  Cayman Chemical 331332.5 MG
Encompass_Preferred
A polyene antibiotic; induces lysis and degradation of B. subtilis as a component of Streptomyces Mg1 extract
Catalog No. 50-294-7453