Learn More
Medchemexpress LLC Lincomycin hydrochloride | 859-18-7 | 99.3% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB0417A100MG
Encompass_Preferred
Lincomycin hydrochloride is the salt form of the antibiotic lincomycin. It affects the synthesis of protein, RNA, and DNA in the mycelium of Act. Roseolus. It inhibits protein synthesis in gram-positive and, to a lesser degree, in gram-negative bacteria by binding to the 50S subunits of bacterial ribosomes and acting on aminoacyl-tRNA binding and the transpeptidation reaction.
  • Salt form of the antibiotic lincomycin
  • Affects the synthesis of protein, RNA, and DNA
  • Inhibits protein synthesis in gram-positive and gram-negative bacteria
  • Binds to the 50S subunits of bacterial ribosomes
  • Acts on aminoacyl-tRNA binding and the transpeptidation reaction
Catalog No. 50-004-06541