Learn More
Medchemexpress LLC HY-16643 10mg , Linerixibat CAS:1345982-69-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1664310MG
Encompass_Preferred
HY-16643 10mg Linerixibat CAS: 1345982-69-5 Linerixibat (GSK2330672) is a highly potent, nonabsorbable and orally active apical sodium-dependent bile acid transporter (ASBT) inhibitor with an IC50 of 42 nM human ASBT. Linerixibat can be used as lipid-lowering agent. Linerixibat has the potential for type 2 diabetes and Primary Biliary Cholangitis treatment. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0191