Learn More
Medchemexpress LLC HY-10394 50mg , Linezolid CAS:165800-03-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1039450MG
Encompass_Preferred
HY-10394 50mg Linezolid CAS: 165800-03-3 Linezolid (PNU-100766) is the first member of the class of oxazolidinone synthetic antibiotic. Linezolid acts by inhibiting the initiation of bacterial protein synthesis. Linezolid is used for the treatment of serious infections caused by Gram-positive bacteria that are resistant to several other antibiotics. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0458