Learn More
Medchemexpress LLC Linezolid-d8 | 1032182-14-1 | 345.40 | 1 MG
SDP
Supplier:  Medchemexpress LLC 1MGHY10394S1
Encompass_Preferred
PNU-100766-d8 is a deuterated and labeled form of Linezolid (HY-10394). Linezolid (PNU-100766) belongs to the oxazolidinone class of synthetic antibiotics. Its mechanism of action involves inhibiting the initiation of bacterial protein synthesis. It is utilized in the treatment of severe infections caused by Gram-positive bacteria that have developed resistance to other antibiotics.
  • Targets antibiotic, bacterial, and isotope-labeled compounds
  • Involved in anti-infection and other pathways
  • Incorporation of stable heavy isotopes of elements like hydrogen, carbon, and others into drug molecules serves primarily as tracers for quantitative analysis during drug development
  • Deuteration is noted for its potential to influence the pharmacokinetic and metabolic characteristics of drugs
  • Inhibits bacterial protein synthesis through its binding to rRNA on both the 30S and 50S ribosomal subunits
Catalog No. 50-003-65400