Learn More
Medchemexpress LLC Linoleamide | 3999-01-7 | 25 MG

Supplier: Medchemexpress LLC 25MGHYN7848
Linoleamide is a linoleic acid amide that regulates Ca (II) ux and inhibits the erg current. This solid, white to off-white compound has a purity of 99.4% and a molecular weight of 279.46, with the chemical formula C18H33NO. It is intended for research use only.
- Regulates Ca (II) ux
- Inhibits the erg current
- Purity of 99.4%
- Molecular weight of 279.46
- Chemical formula C18H33NO
- Appearance: Solid, white to off-white
- Melting point: 57-58°C
- Initial source from microorganisms
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.