Learn More
Medchemexpress LLC Linoleamide | 3999-01-7 | 5 MG

Supplier: Medchemexpress LLC HYN78485MG
Linoleamide is a linoleic acid amide that regulates Ca(II) ux and inhibits the erg current. This product is for research use only and not sold to patients.
- Regulates Ca(II) ux
- Inhibits the erg current
- Purity of 99.4%
- Molecular weight: 279.46
- Formula: C18H33NO
- Appearance: Solid
- Color: White to off-white
- Initial source: Microorganisms
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.