Learn More
Medchemexpress LLC Linoleate sodium | 822-17-3 | 99.9% | 302.43 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYW011398
Encompass_Preferred
Linoleate sodium is an orally active IL8 regulator via the JNK and NF-κB pathway. It can change the composition of fatty acids and the production of metabolites in cells. Linoleate sodium has anti-inflammatory, immune-regulating, and tumor cell growth-affecting activities.
  • Orally active IL8 regulator via the JNK and NF-κB pathway.
  • Changes the composition of fatty acids and the production of metabolites in cells.
  • Possesses anti-inflammatory, immune-regulating, and tumor cell growth-affecting activities.
Catalog No. 50-003-66760