Learn More
Cayman Chemical LInoleIc ACD glycIdyl STR 25mg
SDP
Supplier:  Cayman Chemical 3684025 MG
Encompass_Preferred
An esterified form of linoleic acid; hydrolyzed to the rat-carcinogenic compound glycidol in vivo; has been found in commercial edible oils and infant formulas
Catalog No. 50-290-9324