Learn More
Medchemexpress LLC Linoleic acid-13C18 | 287111-25-5 | 298.31 | 1 MG
Supplier: Medchemexpress LLC HYN0729S21MG
Linoleic acid-13C18 is the 13C labeled Linoleic acid. Linoleic acid is a common polyunsaturated fatty acid (PUFA) found in plant-based oils, nuts, and seeds. It is a part of membrane phospholipids and functions as a structural component to maintain a certain level of membrane fluidity of the transdermal water barrier of the epidermis. Linoleic acid also induces red blood cells and hemoglobin damage via oxidative mechanisms. It is for research use only.
- Can be used as a tracer in research.
- Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
- Stable heavy isotopes are incorporated into drug molecules for quantitation during drug development.
- Purity: 99.10%.