Learn More
Medchemexpress LLC Linoleic acid alkyne | 1219038-31-9 | 96.1% | C18H28O2 | 100 UG
Supplier: Medchemexpress LLC HY145469100UG
Linoleic acid alkyne is an ω-alkyne derivative of Linoleic acid. It can be used to synthesize other alkyne-containing products, such as glycerophospholipids, for click chemistry.
- ω-alkyne derivative of linoleic acid
- Synthesizes other alkyne-containing products
- Synthesizes glycerophospholipids
- Suitable for click chemistry
- For research use only