Learn More
Cayman Chemical LInoleIc AcId-d11 1mg
SDP
Supplier:  Cayman Chemical 90021931 MG
Encompass_Preferred
An internal standard for the quantification of linoleic acid by GC- or LC-MS
Catalog No. 50-289-2664