Learn More
Cayman Chemical LInoleIc AcId-d4 1mg
SDP
Supplier:  Cayman Chemical 3901501 MG
Encompass_Preferred
An internal standard for the quantification of linoleic acid by GC- or LC-MS
Catalog No. 50-291-8824