Learn More
Medchemexpress LLC Linoleic acid-d5 | 1169764-58-2 | 285.48 | 500 UG
SDP
Supplier:  Medchemexpress LLC HYN0729S4500UG
Encompass_Preferred
Linoleic Acid-d5 is the deuterium labeled Linoleic acid. Linoleic acid is a common polyunsaturated fatty acid (PUFA) found in plant-based oils, nuts, and seeds. It functions as a structural component in membrane phospholipids, maintaining membrane fluidity of the transdermal water barrier of the epidermis. This compound can induce red blood cell and hemoglobin damage via an oxidative mechanism. It is for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, primarily as tracers for quantitation during the drug development process
  • Deuteration has garnered attention due to its potential to influence the pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-15807